Assay Details
Target Gene Details
Entrez Gene ID: | 54394 |
Gene Name: | cytokine receptor-like factor 3 |
Gene Aliases: |
BB164954, Creme9, Cytor4 |
Location: |
Chr.11:80046493-80080991 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crlf3 | NM_001277106.1 | NP_001264035.1 | ||
NM_018776.2 | NP_061246.1 | |||
AF046060.1 | AAD02423.1 | |||
AF120152.1 | AAD31759.1 | |||
AK037880.1 | BAC29888.1 | |||
AK078569.1 | ||||
AK137528.1 | BAE23396.1 | |||
AK146798.1 | BAE27440.1 | |||
AK178399.1 | ||||
AK197632.1 | ||||
BC004780.1 | AAH04780.1 |
More Information
Set Membership: |
Intragenic Non-exonic |