Assay Details
Target Gene Details
Entrez Gene ID: | 69131 |
Gene Name: | cyclin-dependent kinase 12 |
Gene Aliases: |
1810022J16Rik, AI646528, Crk7, Crkrs, D11Ertd752e, Pksc |
Location: |
Chr.11:98202326-98281101 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdk12 | NM_001109626.1 | NP_001103096.1 | ||
NM_001109628.1 | NP_001103098.1 | |||
NM_026952.2 | NP_081228.2 | |||
AK049376.1 | ||||
AK129237.1 | BAC98047.1 | |||
AK137734.1 | ||||
AY072295.1 | AAL69526.1 | |||
BC116645.1 |
More Information
Set Membership: |
Intragenic Non-exonic |