Assay Details
Target Gene Details
Entrez Gene ID: | 16834 |
Gene Name: | component of oligomeric golgi complex 1 |
Gene Aliases: |
6030445I15, Ldlb, mKIAA1381 |
Location: |
Chr.11:113649178-113669331 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cog1 | NM_013581.3 | NP_038609.3 | ||
AF109377.2 | AAD13780.2 | |||
AK031510.1 | ||||
AK034636.2 | BAE43288.1 | |||
AK152464.1 | BAE31241.1 | |||
AK158507.1 | BAE34539.1 | |||
AK159497.1 | BAE35130.1 | |||
AK162971.1 | BAE37141.1 | |||
AK198325.1 | ||||
AK207324.1 | ||||
AK214519.1 | ||||
AK218844.1 | ||||
AK219755.1 | ||||
AK220387.1 | BAD90252.1 | |||
BC049541.1 | AAH49541.1 | |||
BC063056.1 | AAH63056.1 |
Target Gene Details
Entrez Gene ID: | 28081 |
Gene Name: | family with sequence similarity 104, member A |
Gene Aliases: |
D11Wsu99e |
Location: |
Chr.11:113661319-113684186 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam104a | NM_138598.5 | 3 | 1714 | NP_613064.2 |
AK009763.2 | 3 | 1596 | BAB26487.1 | |
AK087540.1 | 3 | 1625 | BAC39919.1 | |
AK145119.1 | 3 | 1534 | ||
AK158872.1 | 3 | 1760 | BAE34705.1 | |
AK159228.1 | 3 | 1613 | BAE34914.1 | |
AK159659.1 | 3 | 1613 | BAE35268.1 | |
AK182299.1 | 1 | 251 | ||
AK186062.1 | ||||
AK192770.1 | ||||
AK204410.1 |