Assay Details
Target Gene Details
Entrez Gene ID: | 20304 |
Gene Name: | chemokine (C-C motif) ligand 5 |
Gene Aliases: |
MuRantes, RANTES, SISd, Scya5, TCP228 |
Location: |
Chr.11:83525778-83530518 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl5 | NM_013653.3 | NP_038681.2 | ||
AF065944.1 | AAC17511.1 | |||
AF065945.1 | AAC17512.1 | |||
AF065946.1 | AAC17513.1 | |||
AF065947.1 | AAC17514.1 | |||
AF128187.1 | AAF22528.1 | |||
AK003101.1 | BAB22566.1 | |||
AK158074.1 | ||||
AY722103.1 | AAU43779.1 | |||
BC033508.1 | AAH33508.1 | |||
CT010315.1 | CAJ18523.1 | |||
M77747.1 | AAA40029.1 | |||
S37648.1 | AAB22330.1 |
More Information
Set Membership: |
Intragenic Non-exonic |