Assay Details
Target Gene Details
Entrez Gene ID: | 20302 |
Gene Name: | chemokine (C-C motif) ligand 3 |
Gene Aliases: |
AI323804, G0S19-1, LD78alpha, MIP-1alpha, MIP1-(a), MIP1-alpha, Mip1a, Scya3 |
Location: |
Chr.11:83647843-83649378 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl3 | NM_011337.2 | 1 | 131 | NP_035467.1 |
AA895994.1 | 1 | 131 | ||
AF065939.1 | 1 | 56 | AAC17506.1 | |
AF065940.1 | 1 | 56 | AAC17507.1 | |
AF065941.1 | 1 | 56 | AAC17508.1 | |
AF065942.1 | 1 | 56 | AAC17509.1 | |
AF065943.1 | 1 | 56 | AAC17510.1 | |
AK137525.1 | 1 | 72 | ||
AK150590.1 | 1 | 111 | BAE29684.1 | |
AK150634.1 | 1 | 111 | BAE29723.1 | |
AK150698.1 | 1 | 111 | BAE29777.1 | |
AK151581.1 | 1 | 111 | BAE30521.1 | |
AK152648.1 | 1 | 111 | BAE31386.1 | |
AK153155.1 | BAE31765.1 | |||
AK155058.1 | 1 | 111 | ||
BC111443.1 | 1 | 55 | ||
EU366956.1 | ||||
J04491.1 | 1 | 77 | AAA40304.1 | |
M23447.1 | 1 | 108 | AAA40146.1 | |
X12531.1 | 1 | 100 | CAA31047.1 |
Target Gene Details
Entrez Gene ID: | 20303 |
Gene Name: | chemokine (C-C motif) ligand 4 |
Gene Aliases: |
AT744.1, Act-2, MIP-1B, Mip1b, Scya4 |
Location: |
Chr.11:83648624-83664683 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl4 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |