Assay Details
Target Gene Details
Entrez Gene ID: | 12418 |
Gene Name: | chromobox 4 |
Gene Aliases: |
MPc2, PC2 |
Location: |
Chr.11:119077571-119086237 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cbx4 | NM_007625.2 | NP_031651.2 | ||
BC045162.1 | ||||
BC117801.1 | ||||
U63387.1 | AAB96874.1 |
More Information
Set Membership: |
Intragenic Non-exonic |