Assay Details
Target Gene Details
Entrez Gene ID: | 14584 |
Gene Name: | glutamine fructose-6-phosphate transaminase 2 |
Gene Aliases: |
AI480523, GFAT2 |
Location: |
Chr.11:49794155-49838620 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfpt2 | NM_013529.3 | 18 | 1969 | NP_038557.1 |
AB016780.1 | 18 | 1935 | BAA74729.1 | |
AK132821.1 | 17 | 1815 | BAE21379.1 | |
BC031928.1 | 18 | 1937 | AAH31928.1 |
More Information
Set Membership: |
Intragenic Exonic |