Assay Details
Target Gene Details
Entrez Gene ID: | 74203 |
Gene Name: | eukaryotic translation initiation factor 4E nuclear import factor 1 |
Gene Aliases: |
2610509L04Rik, A930019J01Rik, AA410001, AU021239, Clast4, D11Ertd166e |
Location: |
Chr.11:3201598-3244588 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif4enif1 | NM_001166547.1 | NP_001160019.1 | ||
NM_001166548.1 | NP_001160020.1 | |||
NM_001166549.1 | NP_001160021.1 | |||
NM_023743.2 | NP_076232.2 | |||
AB031388.1 | BAB11963.1 | |||
AK041598.1 | BAC30999.1 | |||
AK088425.1 | BAC40347.1 | |||
AK128990.1 | ||||
AK162367.1 | BAE36876.1 | |||
AK165678.1 | ||||
AK205705.1 | ||||
BC033410.1 | AAH33410.1 |
More Information
Set Membership: |
Intragenic Non-exonic |