Assay Details
Target Gene Details
Entrez Gene ID: | 217207 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 8 |
Gene Aliases: |
Ddx8, mDEAH6, mKIAA4096 |
Location: |
Chr.11:101732956-101767358 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhx8 | NM_144831.2 | NP_659080.2 | ||
AK078552.1 | ||||
AK142226.1 | BAE24985.1 | |||
AK170616.1 | ||||
AK172600.1 | ||||
AK220485.1 | BAD90286.1 | |||
BC022656.1 | AAH22656.1 | |||
BC132445.1 |
More Information
Set Membership: |
Intragenic Non-exonic |