Assay Details
Target Gene Details
Entrez Gene ID: | 12416 |
Gene Name: | chromobox 2 |
Gene Aliases: |
M33, MOD2, pc |
Location: |
Chr.11:119023019-119031274 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cbx2 | NM_007623.2 | NP_031649.2 | ||
AK157869.1 | BAE34242.1 | |||
BC035199.1 | AAH35199.1 | |||
X62537.1 | CAA44398.1 | |||
Y13274.1 | CAA73723.1 |