Assay Details
Target Gene Details
Entrez Gene ID: | 14017 |
Gene Name: | ecotropic viral integration site 2a |
Gene Aliases: |
AW491894, Evi-2, Evi2 |
Location: |
Chr.11:79526560-79530609 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Evi2a | NM_001033711.1 | 1 | 46 | NP_001028883.1 |
NM_010161.3 | 1 | 46 | NP_034291.1 | |
AK005353.1 | ||||
AK046407.1 | BAC32711.1 | |||
AK170527.1 | BAE41859.1 | |||
BC038124.1 | AAH38124.1 | |||
M34896.1 | AAA37583.1 |
Target Gene Details
Entrez Gene ID: | 101488212 |
Gene Name: | predicted gene 21975 |
Gene Aliases: |
Evi2a-evi2b |
Location: |
Chr.11:79513385-79530609 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm21975 | NM_146023.4 | 1 | 46 | NP_666135.1 |
BB626486.1 |
Target Gene Details
Entrez Gene ID: | 18015 |
Gene Name: | neurofibromatosis 1 |
Gene Aliases: |
AW494271, E030030H24Rik, Nf-1 |
Location: |
Chr.11:79335988-79581609 on Build GRCm38 |
Assay Gene Location: | Within Intron 36 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nf1 | NM_010897.2 | NP_035027.1 | ||
L10370.1 | AAA68132.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |