Assay Details
Target Gene Details
Entrez Gene ID: | 11465 |
Gene Name: | actin, gamma, cytoplasmic 1 |
Gene Aliases: |
AL023024, Actg, Actl, E51 |
Location: |
Chr.11:120345689-120348491 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Actg1 | NM_009609.2 | NP_033739.1 | ||
AF195094.1 | AAF08293.1 | |||
AK012251.1 | ||||
AK076081.1 | BAC36167.1 | |||
AK087983.1 | BAC40075.1 | |||
AK149927.1 | BAE29172.1 | |||
AK150209.1 | BAE29380.1 | |||
AK152202.1 | BAE31031.1 | |||
AK153446.1 | BAE32001.1 | |||
AK157726.1 | BAE34171.1 | |||
AK159867.1 | BAE35441.1 | |||
AK162157.1 | BAE36758.1 | |||
BC003337.1 | AAH03337.1 | |||
BC021796.1 | AAH21796.1 | |||
BC023248.1 | AAH23248.1 | |||
BC099371.1 | AAH99371.1 | |||
M21495.1 | AAA37168.1 | |||
X13055.1 | CAA31455.1 |
More Information
Set Membership: |
Intragenic Non-exonic |