Assay Details
Target Gene Details
Entrez Gene ID: | 66884 |
Gene Name: | amyloid beta precursor protein (cytoplasmic tail) binding protein 2 |
Gene Aliases: |
1300003O07Rik, AI465480, PAT1 |
Location: |
Chr.11:85191308-85235120 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Appbp2 | NM_025825.3 | 8 | 1195 | NP_080101.1 |
AK004888.1 | 8 | 1206 | ||
AK005440.1 | 8 | 901 | BAB24034.1 | |
AK133853.1 | 8 | 1187 | ||
BC018442.1 | 8 | 1173 | AAH18442.1 | |
BC019152.1 | 8 | 1196 | AAH19152.1 |