Assay Details
Target Gene Details
Entrez Gene ID: | 268390 |
Gene Name: | AHA1, activator of heat shock protein ATPase 2 |
Gene Aliases: |
- |
Location: |
Chr.11:23487882-23498039 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ahsa2 | NM_001290654.1 | 2 | 399 | NP_001277583.1 |
NM_001290655.1 | 2 | 404 | NP_001277584.1 | |
NM_172391.4 | 2 | 404 | NP_765979.3 | |
AK033696.1 | 2 | 312 | ||
AK035941.1 | 2 | 177 | BAC29251.1 | |
AK093945.1 | 2 | 311 | BAC04256.1 | |
AK137116.1 | 2 | 315 | BAE23240.1 | |
AK154196.1 | 2 | 315 | BAE32431.1 | |
AK155265.1 | 2 | 315 | BAE33153.1 | |
AK186357.1 | 2 | 157 | ||
BC038397.1 | 2 | 290 | ||
BC052829.1 | 2 | 322 | AAH52829.1 | |
BC064012.1 | 2 | 224 | AAH64012.1 | |
BY013732.1 | 2 | 309 | ||
BY023007.1 | 2 | 288 | ||
CJ147821.1 | 2 | 405 |
More Information
Set Membership: |
Intragenic Exonic |