Assay Details
Target Gene Details
Entrez Gene ID: | 104184 |
Gene Name: | bleomycin hydrolase |
Gene Aliases: |
AI035728, Bh, Bmh |
Location: |
Chr.11:76945656-76987389 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Blmh | NM_178645.4 | NP_848760.1 | ||
AK029274.1 | ||||
AK035228.1 | BAC28989.1 | |||
AK040799.1 | BAC30706.1 | |||
AK049461.1 | ||||
AK051441.1 | BAC34639.1 | |||
AK053872.1 | BAC35568.1 | |||
AK150438.1 | BAE29560.1 | |||
AK150896.1 | ||||
AK167325.1 | BAE39427.1 | |||
BC024090.1 | AAH24090.1 | |||
BC027037.1 | AAH27037.1 | |||
BC027362.1 | AAH27362.1 | |||
BC027403.1 | AAH27403.1 |
More Information
Set Membership: |
Intragenic Non-exonic |