Assay Details
Target Gene Details
Entrez Gene ID: | 11818 |
Gene Name: | apolipoprotein H |
Gene Aliases: |
B2GPI, beta-2-GPI, beta2-GPI |
Location: |
Chr.11:108395297-108414396 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apoh | NM_013475.4 | 6 | 743 | NP_038503.4 |
AA273732.1 | 3 | 358 | ||
AK010926.1 | 6 | 731 | BAB27271.1 | |
BC019811.1 | 6 | 731 | AAH19811.1 | |
BC053338.1 | 6 | 719 | AAH53338.1 | |
BU756628.1 | 3 | 460 | ||
D10056.1 | 6 | 717 | BAA00945.1 | |
S70439.1 | 6 | 727 | AAB30789.1 | |
Y11356.1 | 6 | 747 | CAA72190.1 |