Assay Details
Target Gene Details
Entrez Gene ID: | 12330 |
Gene Name: | calnexin |
Gene Aliases: |
1110069N15Rik, AI988026, Cnx, D11Ertd153e |
Location: |
Chr.11:50293957-50325673 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Canx | NM_001110499.1 | NP_001103969.1 | ||
NM_001110500.1 | NP_001103970.1 | |||
NM_007597.3 | NP_031623.1 | |||
AK028279.1 | ||||
AK084175.1 | BAC39133.1 | |||
AK159298.1 | BAE34971.1 | |||
AK165508.1 | BAE38226.1 | |||
AK166649.1 | BAE38916.1 | |||
BC012408.1 | AAH12408.1 | |||
BC040244.1 | AAH40244.1 | |||
L18888.1 | AAA21014.1 | |||
L23865.1 | AAA62450.1 |
More Information
Set Membership: |
Intragenic Non-exonic |