Assay Details
Target Gene Details
Entrez Gene ID: | 192650 |
Gene Name: | calcium binding protein 7 |
Gene Aliases: |
- |
Location: |
Chr.11:4738821-4746778 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cabp7 | NM_138948.3 | NP_620398.1 | ||
AF419324.1 | AAL14997.1 | |||
BC104356.1 | AAI04357.1 | |||
BC104357.1 | AAI04358.1 |