Assay Details
Target Gene Details
Entrez Gene ID: | 12029 |
Gene Name: | B cell CLL/lymphoma 6, member B |
Gene Aliases: |
Bazf |
Location: |
Chr.11:70224127-70229798 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcl6b | NM_007528.3 | NP_031554.1 | ||
AB011665.1 | BAA31223.1 | |||
AK032647.1 | BAC27970.1 | |||
AK156733.1 | BAE33827.1 | |||
BC090998.1 | AAH90998.1 |
More Information
Set Membership: |
Intragenic Non-exonic |