Assay Details
Target Gene Details
Entrez Gene ID: | 12029 |
Gene Name: | B cell CLL/lymphoma 6, member B |
Gene Aliases: |
Bazf |
Location: |
Chr.11:70224127-70229798 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcl6b | NM_007528.3 | 6 | 1096 | NP_031554.1 |
AB011665.1 | 5 | 1019 | BAA31223.1 | |
AK032647.1 | 1 | 87 | BAC27970.1 | |
AK156733.1 | 6 | 1058 | BAE33827.1 | |
BC090998.1 | 6 | 1056 | AAH90998.1 |
More Information
Set Membership: |
Intragenic Exonic |