Assay Details
Target Gene Details
Entrez Gene ID: | 66358 |
Gene Name: | ADP-ribose/CDP-alcohol diphosphatase, manganese dependent |
Gene Aliases: |
2310004I24Rik, MDS006 |
Location: |
Chr.11:67037880-67052623 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adprm | NM_025510.3 | NP_079786.2 | ||
AK009142.1 | BAB26101.1 | |||
AK010520.1 | ||||
AK145941.1 | BAE26771.1 | |||
BC004029.1 | AAH04029.1 | |||
BY118851.1 |