Assay Details
Target Gene Details
Entrez Gene ID: | 11686 |
Gene Name: | arachidonate 12-lipoxygenase, 12R type |
Gene Aliases: |
Aloxe2, e-LOX2 |
Location: |
Chr.11:69157072-69169791 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Alox12b | NM_009659.2 | NP_033789.1 | ||
AF059251.1 | AAC79681.1 | |||
AK019091.1 | BAB31541.1 | |||
BC113149.1 | AAI13150.1 | |||
Y14334.1 | CAA74714.1 |
More Information
Set Membership: |
Intragenic Non-exonic |