Assay Details
Target Gene Details
Entrez Gene ID: | 103742 |
Gene Name: | migration and invasion enhancer 1 |
Gene Aliases: |
1810046J19Rik, AI463380, Rdx12 |
Location: |
Chr.11:98437708-98438988 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mien1 | NM_025559.2 | NP_079835.1 | ||
AK002959.1 | BAB22480.1 | |||
AK007795.1 | BAB25261.1 | |||
AK009922.1 | BAB26586.1 | |||
AK041314.1 | BAC30901.1 | |||
BC021589.1 | AAH21589.1 | |||
CA530220.1 |