Assay Details
Target Gene Details
Entrez Gene ID: | 57316 |
Gene Name: | C1D nuclear receptor co-repressor |
Gene Aliases: |
1110036E10Rik, AI875855, SUN-CoR |
Location: |
Chr.11:17257618-17269176 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1d | NM_020558.3 | NP_065583.2 | ||
AK028702.1 | BAC26075.1 | |||
AK035169.1 | BAC28967.1 | |||
BC005436.1 | AAH05436.1 | |||
X95591.1 | CAA64844.1 |
More Information
Set Membership: |
Intragenic Non-exonic |