Assay Details
Target Gene Details
Entrez Gene ID: | 65257 |
Gene Name: | ankyrin repeat and SOCS box-containing 3 |
Gene Aliases: |
2400011J03Rik, Asb-3 |
Location: |
Chr.11:30954194-31102704 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asb3 | NM_023906.3 | 7 | 1110 | NP_076395.2 |
AF155354.1 | 6 | 925 | AAD38810.1 | |
AK010315.1 | 7 | 1046 | BAB26847.1 | |
AK028489.1 | 8 | 1216 | ||
AK038086.1 | 7 | 1091 | ||
AK143580.1 | 7 | 1045 | BAE25448.1 | |
BC023086.1 | 7 | 1044 | AAH23086.1 |