Assay Details
Target Gene Details
Entrez Gene ID: | 65257 |
Gene Name: | ankyrin repeat and SOCS box-containing 3 |
Gene Aliases: |
2400011J03Rik, Asb-3 |
Location: |
Chr.11:30954194-31102704 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asb3 | NM_023906.3 | 4 | 591 | NP_076395.2 |
AF155354.1 | 3 | 406 | AAD38810.1 | |
AK010315.1 | 4 | 527 | BAB26847.1 | |
AK028489.1 | 5 | 697 | ||
AK038086.1 | 4 | 572 | ||
AK143580.1 | 4 | 526 | BAE25448.1 | |
BB618690.1 | ||||
BC023086.1 | 4 | 525 | AAH23086.1 |