Assay Details
Target Gene Details
Entrez Gene ID: | 66201 |
Gene Name: | Vps20-associated 1 homolog (S. cerevisiae) |
Gene Aliases: |
1110001D18Rik, 1110059P08Rik, AU040813, C85340, LIP5, SBP1 |
Location: |
Chr.10:14654760-14705527 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vta1 | NM_025418.3 | 6 | 597 | NP_079694.2 |
AK002611.1 | 6 | 596 | BAB22228.1 | |
AK003213.1 | 5 | 514 | ||
AK004323.1 | 6 | 669 | BAE43171.1 | |
AK009227.2 | 6 | 586 | BAB26150.1 | |
BC026752.1 | 6 | 590 | AAH26752.1 | |
CX212626.1 | 6 | 598 |
More Information
Set Membership: |
Intragenic Exonic |