Assay Details
Target Gene Details
Entrez Gene ID: | 66201 |
Gene Name: | Vps20-associated 1 homolog (S. cerevisiae) |
Gene Aliases: |
1110001D18Rik, 1110059P08Rik, AU040813, C85340, LIP5, SBP1 |
Location: |
Chr.10:14654760-14705527 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vta1 | NM_025418.3 | NP_079694.2 | ||
AK002611.1 | BAB22228.1 | |||
AK003213.1 | ||||
AK004323.1 | BAE43171.1 | |||
AK009227.2 | BAB26150.1 | |||
AK044986.1 | 1 | 107 | ||
AK170710.1 | 1 | 107 | ||
BC026752.1 | AAH26752.1 | |||
CX212626.1 |
More Information
Set Membership: |
Intragenic Non-exonic |