Assay Details
Target Gene Details
Entrez Gene ID: | 73324 |
Gene Name: | clathrin heavy chain linker domain containing 1 |
Gene Aliases: |
1700034F02Rik |
Location: |
Chr.11:29547950-29578360 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clhc1 | NM_001163521.1 | 6 | 1027 | NP_001156993.1 |
NM_028504.1 | 6 | 1060 | NP_082780.1 | |
AK006591.1 | 6 | 1061 | BAB24663.1 | |
BC137683.1 | 6 | 984 | ||
BC144828.1 | 6 | 951 |
More Information
Set Membership: |
Intragenic Exonic |