Assay Details
Target Gene Details
Entrez Gene ID: | 140721 |
Gene Name: | CASK-interacting protein 2 |
Gene Aliases: |
1600028L06Rik, mKIAA1139 |
Location: |
Chr.11:115799183-115813592 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Caskin2 | NM_080643.2 | NP_542374.2 | ||
AF451978.1 | AAL49759.1 | |||
AK129297.1 | BAC98107.1 | |||
BC053083.1 | AAH53083.1 |
More Information
Set Membership: |
Intragenic Non-exonic |