Assay Details
Target Gene Details
Entrez Gene ID: | 20509 |
Gene Name: | solute carrier family 19 (folate transporter), member 1 |
Gene Aliases: |
AI323572, RFC, RFC-1, RFC1 |
Location: |
Chr.10:77032269-77050432 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc19a1 | NM_001199271.1 | NP_001186200.1 | ||
NM_031196.3 | NP_112473.1 | |||
AK002560.1 | BAB22188.1 | |||
AK050714.1 | 1 | 103 | BAC34390.1 | |
AK079592.1 | BAC37693.1 | |||
AK085024.2 | ||||
AK088803.1 | BAC40582.1 | |||
AK138577.1 | ||||
BC015263.1 | AAH15263.1 | |||
BF786711.1 | ||||
BY791593.1 | ||||
L36539.1 | AAB38483.1 | |||
U32469.1 | AAC52258.1 | |||
U66103.1 | AAC53287.1 |
More Information
Set Membership: |
Intragenic Non-exonic |