Assay Details
Target Gene Details
Entrez Gene ID: | 27375 |
Gene Name: | tight junction protein 3 |
Gene Aliases: |
- |
Location: |
Chr.10:81273188-81291581 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tjp3 | NM_001282095.1 | NP_001269024.1 | ||
NM_001282096.1 | NP_001269025.1 | |||
NM_013769.3 | NP_038797.2 | |||
AF157006.1 | AAF24175.1 | |||
AK029083.1 | BAC26285.1 | |||
AK134100.1 | BAE22013.1 | |||
AK167587.1 | ||||
BC012518.1 | AAH12518.1 |
More Information
Set Membership: |
Intragenic Non-exonic |