Assay Details
Target Gene Details
Entrez Gene ID: | 20503 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 7 |
Gene Aliases: |
4921534N07Rik, Mct2 |
Location: |
Chr.10:125219284-125389562 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc16a7 | NM_011391.1 | NP_035521.1 | ||
AF058054.1 | AAC13719.1 | |||
AK085398.1 | 1 | 491 | ||
DQ140161.1 | 1 | 489 |
More Information
Set Membership: |
Intragenic Non-exonic |