Assay Details
Target Gene Details
Entrez Gene ID: | 20393 |
Gene Name: | serum/glucocorticoid regulated kinase 1 |
Gene Aliases: |
Sgk |
Location: |
Chr.10:21882184-21999903 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sgk1 | NM_001161845.2 | NP_001155317.2 | ||
NM_001161847.2 | NP_001155319.1 | |||
NM_001161848.2 | NP_001155320.1 | |||
NM_001161849.2 | NP_001155321.1 | |||
NM_001161850.2 | NP_001155322.1 | |||
NM_011361.3 | NP_035491.1 | |||
AF139638.1 | AAD43302.1 | |||
AF205855.1 | AAF19429.1 | |||
AK086892.1 | ||||
AK132234.1 | BAE21048.1 | |||
AK146037.1 | BAE26849.1 | |||
AK146062.1 | BAE26871.1 | |||
AK159131.1 | BAE34843.1 | |||
AK167364.1 | BAE39461.1 | |||
BC002222.1 | AAH02222.1 | |||
BC005720.1 | AAH05720.1 | |||
BC070401.1 | AAH70401.1 |
More Information
Set Membership: |
Intragenic Non-exonic |