Assay Details
Target Gene Details
Entrez Gene ID: | 64009 |
Gene Name: | spectrin repeat containing, nuclear envelope 1 |
Gene Aliases: |
8B, A330049M09Rik, BE692247, C130039F11Rik, CPG2, Myne1, mKIAA1756 |
Location: |
Chr.10:5020192-5550829 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 83 - Intron 83 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Syne1 | NM_001079686.1 | NP_001073154.1 | ||
NM_022027.2 | NP_071310.2 | |||
AF281869.1 | AAG24392.1 | |||
AF281870.1 | AAG24393.1 | |||
AK036828.1 | BAC29595.1 | |||
BC041779.1 | AAH41779.1 | |||
BC054456.1 | AAH54456.1 | |||
BC110428.1 | ||||
BC111034.1 | AAI11035.1 |