Assay Details
Target Gene Details
Entrez Gene ID: | 70428 |
Gene Name: | polymerase (RNA) III (DNA directed) polypeptide B |
Gene Aliases: |
2700078H01Rik, A330032P03Rik, C85372, RPC2 |
Location: |
Chr.10:84621456-84727178 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr3b | NM_027423.1 | NP_081699.1 | ||
AK039244.1 | ||||
AK039366.1 | BAC30327.1 | |||
AK165886.1 | BAE38438.1 | |||
BC068143.1 | AAH68143.1 |
More Information
Set Membership: |
Intragenic Non-exonic |