Assay Details
Target Gene Details
Entrez Gene ID: | 64009 |
Gene Name: | spectrin repeat containing, nuclear envelope 1 |
Gene Aliases: |
8B, A330049M09Rik, BE692247, C130039F11Rik, CPG2, Myne1, mKIAA1756 |
Location: |
Chr.10:5020192-5550829 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 70 - Intron 70 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Syne1 | NM_001079686.1 | NP_001073154.1 | ||
AF281870.1 | AAG24393.1 |