Assay Details
Target Gene Details
Entrez Gene ID: | 20587 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 |
Gene Aliases: |
AU020204, Baf47, Ini1, SNF5/INI1, Snf5 |
Location: |
Chr.10:75896769-75921614 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarcb1 | NM_001161853.1 | NP_001155325.1 | ||
NM_011418.2 | NP_035548.1 | |||
AB017343.1 | BAB12427.1 | |||
AB017344.1 | BAB12428.1 | |||
AB041578.1 | BAA95062.1 | |||
AJ011739.1 | CAA09760.1 | |||
AJ011740.1 | CAA09761.1 | |||
AK008558.1 | ||||
AK075906.1 | BAC36045.1 | |||
AK145552.1 | BAE26504.1 | |||
AK150169.1 | BAE29357.1 | |||
BC025163.1 | AAH25163.1 |
More Information
Set Membership: |
Intragenic Non-exonic |