Assay Details
Target Gene Details
Entrez Gene ID: | 73106 |
Gene Name: | protease, serine 57 |
Gene Aliases: |
2900092M14Rik, GLGL782, Prssl1, UNQ782 |
Location: |
Chr.10:79781474-79795605 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss57 | NM_001042710.1 | NP_001036175.1 | ||
AK153740.1 | ||||
AK158376.1 | ||||
BC116380.2 |
More Information
Set Membership: |
Intragenic Non-exonic |