Assay Details
Target Gene Details
Entrez Gene ID: | 56535 |
Gene Name: | peroxisomal biogenesis factor 3 |
Gene Aliases: |
1700014F15Rik, 2810027F19Rik, 2900010N04Rik |
Location: |
Chr.10:13523842-13553149 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pex3 | NM_001164195.1 | NP_001157667.1 | ||
NM_019961.3 | NP_064345.1 | |||
AF152996.1 | AAF14524.1 | |||
AK005315.1 | ||||
AK005978.1 | ||||
AK012827.1 | ||||
AK075820.1 | BAC35986.1 | |||
AK146934.1 | BAE27544.1 | |||
BC033415.1 | AAH33415.1 | |||
BC037606.1 | AAH37606.1 |