Assay Details
Target Gene Details
Entrez Gene ID: | 56535 |
Gene Name: | peroxisomal biogenesis factor 3 |
Gene Aliases: |
1700014F15Rik, 2810027F19Rik, 2900010N04Rik |
Location: |
Chr.10:13523842-13553149 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pex3 | NM_001164195.1 | 11 | 1239 | NP_001157667.1 |
NM_019961.3 | 11 | 1239 | NP_064345.1 | |
AF152996.1 | 11 | 1162 | AAF14524.1 | |
AK005315.1 | 11 | 1095 | ||
AK005978.1 | 10 | 936 | ||
AK012827.1 | 11 | 1217 | ||
AK075820.1 | 11 | 1154 | BAC35986.1 | |
AK146934.1 | 11 | 1218 | BAE27544.1 | |
AV256241.1 | 2 | 99 | ||
BC033415.1 | 11 | 1121 | AAH33415.1 | |
BC037606.1 | 11 | 1202 | AAH37606.1 |