Assay Details
Target Gene Details
Entrez Gene ID: | 56214 |
Gene Name: | secretory carrier membrane protein 4 |
Gene Aliases: |
2410022D05Rik, AI448940, AU022392, AW742369, Sc4 |
Location: |
Chr.10:80602882-80615783 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Scamp4 | NM_019575.4 | NP_062521.1 | ||
AF224721.1 | AAF35368.1 | |||
AF295102.1 | AAG02502.1 | |||
AK019112.1 | ||||
AK078226.1 | ||||
AK154094.1 | BAE32371.1 | |||
AK160755.1 | BAE35991.1 | |||
AK168554.1 | BAE40428.1 | |||
BC018215.1 | AAH18215.1 | |||
BC054104.2 | AAH54104.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |