Assay Details
Target Gene Details
Entrez Gene ID: | 56533 |
Gene Name: | regulator of G-protein signaling 17 |
Gene Aliases: |
6430507P11Rik, Rgsz2 |
Location: |
Chr.10:5825663-5922400 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs17 | NM_001161822.1 | 5 | 510 | NP_001155294.1 |
NM_019958.4 | 4 | 440 | NP_064342.1 | |
AF191555.1 | 4 | 382 | AAF05758.1 | |
AK018279.1 | 4 | 424 | BAB31145.1 | |
AK030492.1 | 4 | 441 | BAC26989.1 | |
AK045770.1 | 5 | 477 | BAC32486.1 | |
AK046009.1 | 4 | 399 | BAC32571.1 | |
AK078745.1 | 4 | 399 | BAC37375.1 | |
AK163188.1 | 4 | 397 | BAE37226.1 | |
BC064782.1 | 4 | 368 | AAH64782.1 |
More Information
Set Membership: |
Intragenic Exonic |