Assay Details
Target Gene Details
Entrez Gene ID: | 338359 |
Gene Name: | suppressor of var1, 3-like 1 (S. cerevisiae) |
Gene Aliases: |
6330443E10Rik |
Location: |
Chr.10:62429329-62451294 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Supv3l1 | NM_181423.2 | NP_852088.1 | ||
AJ968954.1 | CAI92124.1 | |||
AK031911.1 | ||||
AK143755.1 | ||||
AK188958.1 | ||||
BC049796.1 | AAH49796.1 |
More Information
Set Membership: |
Intragenic Non-exonic |