Assay Details
Target Gene Details
Entrez Gene ID: | 216233 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
8030460M17, AI527257, AW108012, CIS2, Cish2, D130043N08Rik, JAB, SOCS-2, SSI-2, hg |
Location: |
Chr.10:95383903-95417253 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Socs2 | NM_007706.4 | NP_031732.1 | ||
AK137403.1 | BAE23343.1 | |||
AK144810.1 | ||||
AK160537.1 | BAE35854.1 | |||
AK162591.1 | BAE36978.1 | |||
BC106153.1 | AAI06154.1 | |||
BY069559.1 |
More Information
Set Membership: |
Intragenic Non-exonic |