Assay Details
Target Gene Details
Entrez Gene ID: | 20397 |
Gene Name: | sphingosine phosphate lyase 1 |
Gene Aliases: |
AI428538, D10Xrf456, S1PL, Spl |
Location: |
Chr.10:61098642-61147686 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sgpl1 | NM_009163.3 | NP_033189.2 | ||
AF036894.1 | AAC03768.1 | |||
AK036747.1 | BAC29562.1 | |||
AK037789.1 | BAC29872.1 | |||
AK043024.1 | BAC31437.1 | |||
AK049342.1 | BAC33695.1 | |||
AK150053.1 | BAE29269.1 | |||
AK173137.1 | BAD32415.1 | |||
BC026135.1 | AAH26135.1 | |||
CT010214.1 | CAJ18422.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |