Assay Details
Target Gene Details
Entrez Gene ID: | 57319 |
Gene Name: | sphingomyelin phosphodiesterase, acid-like 3A |
Gene Aliases: |
0610010C24Rik, AI529588, ASM3A, ASML3, ASML3A |
Location: |
Chr.10:57794544-57811830 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smpdl3a | NM_020561.2 | 4 | 641 | NP_065586.3 |
AK002455.1 | 4 | 580 | ||
AK151683.1 | 4 | 527 | BAE30607.1 | |
AK152282.1 | 4 | 542 | BAE31096.1 | |
AK185060.1 | 3 | 387 | ||
AK194580.1 | ||||
AK196251.1 | 2 | 113 | ||
AK205531.1 | ||||
BC139288.1 | 4 | 533 | ||
BC139289.1 | 4 | 533 | ||
Y08135.1 | 4 | 595 | CAA69329.1 |
More Information
Set Membership: |
Intragenic Exonic |