Assay Details
Target Gene Details
Entrez Gene ID: | 71137 |
Gene Name: | regulatory factor X, 4 (influences HLA class II expression) |
Gene Aliases: |
4933412G19Rik, NYD-sp10 |
Location: |
Chr.10:84756048-84906538 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rfx4 | NM_001024918.1 | 4 | 681 | NP_001020089.1 |
AB086957.1 | 4 | 535 | BAD07408.1 | |
AK034131.1 | 4 | 668 | BAC28598.1 | |
AY102010.2 | 4 | 564 | AAM52485.2 | |
AY342003.1 | 4 | 546 | AAQ17042.1 | |
GQ496603.1 | ||||
GQ496604.1 |
More Information
Set Membership: |
Intragenic Exonic |