Assay Details
Target Gene Details
Entrez Gene ID: | 414101 |
Gene Name: | RIKEN cDNA E130317F20 gene |
Gene Aliases: |
- |
Location: |
Chr.10:79851377-79854971 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
E130317F20Rik | NR_029447.1 | 1 | 315 | |
AK148071.1 | 1 | 316 | ||
AK148204.1 |
Target Gene Details
Entrez Gene ID: | 19205 |
Gene Name: | polypyrimidine tract binding protein 1 |
Gene Aliases: |
AA407203, AL033359, HNRPI, PTB-1, PTB2, PTB3, PTB4, Ptb, pPTB |
Location: |
Chr.10:79854432-79864435 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptbp1 | NM_001077363.2 | 1 | 226 | NP_001070831.1 |
NM_001283013.1 | 1 | 226 | NP_001269942.1 | |
NM_008956.3 | 1 | 226 | NP_032982.2 | |
AK033319.1 | BAC28230.1 | |||
AK036745.1 | BAC29560.1 | |||
AK037607.1 | ||||
AK041143.1 | BAC30837.1 | |||
AK043824.1 | BAC31665.1 | |||
AK044956.1 | BAC32158.1 | |||
AK050498.1 | BAC34292.1 | |||
AK051669.1 | BAC34712.1 | |||
AK053926.1 | ||||
AK082600.1 | BAC38544.1 | |||
AK088126.1 | BAC40161.1 | |||
AK088483.1 | BAC40383.1 | |||
AK149941.1 | BAE29183.1 | |||
AK150840.1 | BAE29899.1 | |||
AK151292.1 | 1 | 54 | BAE30277.1 | |
AK153561.1 | 1 | 54 | BAE32096.1 | |
AK163274.1 | 1 | 51 | BAE37269.1 | |
AK168359.1 | BAE40295.1 | |||
AK168392.1 | ||||
AK172712.1 | BAE43140.1 | |||
BC007472.1 | 1 | 43 | AAH07472.1 | |
BC028848.1 | AAH28848.1 | |||
BC066210.1 | AAH66210.1 | |||
BC086489.1 | AAH86489.1 | |||
BY167466.1 | 1 | 54 | ||
CJ075278.1 | 1 | 232 | ||
X52101.1 | CAA36321.1 |