Assay Details
Target Gene Details
Entrez Gene ID: | 67418 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 4 |
Gene Aliases: |
3732410E19Rik, 3830425H19Rik, AI788954, AW146233, PPIase |
Location: |
Chr.10:7792891-7822563 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ppil4 | NM_026141.3 | NP_080417.2 | ||
AK011443.1 | BAB27623.1 | |||
AK012371.1 | BAB28194.1 | |||
AK014453.1 | ||||
AK017370.1 | BAB30711.1 | |||
AK049634.1 | ||||
AK164127.1 | ||||
AK167168.1 | ||||
AK167258.1 | BAE39374.1 | |||
BC004652.1 | AAH04652.1 | |||
BC079912.1 | AAH79912.1 |
More Information
Set Membership: |
Intragenic Non-exonic |